ENAH5802DA3F Enamine

5-(2,3-dihydro-1-benzofuran-5-yl)-1,2-oxazole-3-carbonyl chloride

  • MDL number MFCD31924204
  • Cas Number : 2344680-43-7
  • Molecular Weight : 249.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7353227

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7353227
Availability details for
Quantity
  • Availability
  • Add to Cart