ENAH5802DAB5 Enamine

dimethyl(2-methylbut-3-yn-2-yl)amine

  • MDL number MFCD25960657
  • Cas Number : 19788-24-0
  • Molecular Weight : 111.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 97-98
Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6943659

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P310,P332+P313,P362,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6943659
pictograms GHS05
hstatements H314,H315
Availability details for
Quantity
  • Availability
  • Add to Cart