ENAH5802DAB8 Enamine

4H,5H,6H,7H-pyrazolo[1,5-a]pyrazin-3-ol dihydrochloride

  • MDL number MFCD31924434
  • Cas Number : 2344685-66-9
  • Molecular Weight : 212.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7355835

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7355835
Availability details for
Quantity
  • Availability
  • Add to Cart