ENAH5802DACB Enamine

[amino(prop-2-yn-1-ylsulfanyl)methylidene]azanium bromide

  • Cas Number : 13702-02-8
  • Molecular Weight : 195.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Br-
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-378642

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-378642
Availability details for
Quantity
  • Availability
  • Add to Cart