ENAH5802DB01 Enamine

2-(4-methoxypyridazin-3-yl)ethan-1-ol

  • MDL number MFCD31924333
  • Cas Number : 2344681-34-9
  • Molecular Weight : 154.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7355653

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7355653
Availability details for
Quantity
  • Availability
  • Add to Cart