ENAH5802DB5A Enamine

methyl 2-(hydroxymethyl)-3,6-dihydro-2H-pyran-2-carboxylate

  • MDL number MFCD32066309
  • Cas Number : 2361634-99-1
  • Molecular Weight : 172.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1704136

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1704136
Availability details for
Quantity
  • Availability
  • Add to Cart