ENAH5802DB97 Enamine

[3-(hydroxymethyl)phenyl](imino)methyl-lambda6-sulfanone

  • MDL number MFCD31924695
  • Cas Number : 2344680-52-8
  • Molecular Weight : 185.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7425894

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7425894
Availability details for
Quantity
  • Availability
  • Add to Cart