ENAH5802DBC5 Enamine

2,4-dichloro-6-fluoro-3-methylphenol

  • MDL number MFCD31924047
  • Cas Number : 2344680-94-8
  • Molecular Weight : 195.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7426425

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7426425
Availability details for
Quantity
  • Availability
  • Add to Cart