ENAH5802DC07 Enamine

[5-chloro-2-(2-methylpropoxy)phenyl]methanol

  • MDL number MFCD11180633
  • Cas Number : 23426-37-1
  • Molecular Weight : 214.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7432173

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7432173
Availability details for
Quantity
  • Availability
  • Add to Cart