ENAH5802DC33 Enamine

4-(chloromethyl)-2-phenyl-4,5-dihydro-1,3-thiazol-4-ol

  • MDL number MFCD29730780
  • Cas Number : 37128-19-1
  • Molecular Weight : 227.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7425442

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7425442
Availability details for
Quantity
  • Availability
  • Add to Cart