ENAH5802DC36 Enamine

N-[(4-hydroxyphenyl)methyl]pyridine-4-carboxamide

  • MDL number MFCD27667133
  • Cas Number : 1710192-79-2
  • Molecular Weight : 228.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7432416

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7432416
Availability details for
Quantity
  • Availability
  • Add to Cart