ENAH5802DC3E Enamine

1-(5-bromopyridin-2-yl)-2-methylpropan-1-ol

  • MDL number MFCD26852465
  • Cas Number : 890150-88-6
  • Molecular Weight : 230.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-5104131

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-5104131
Availability details for
Quantity
  • Availability
  • Add to Cart