ENAH5802DC65 Enamine

{2-[3-(trifluoromethyl)phenyl]-1,3-oxazol-4-yl}methanol

  • MDL number MFCD32069425
  • Cas Number : 1448866-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60-63
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7435524

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7435524
Availability details for
Quantity
  • Availability
  • Add to Cart