ENAH5802DC68 Enamine

N-(5-amino-3-methylpyridin-2-yl)-3-hydroxybenzamide

  • MDL number MFCD26241382
  • Cas Number : 1707579-55-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7444660

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7444660
Availability details for
Quantity
  • Availability
  • Add to Cart