ENAH5802DC6B Enamine

6-chloro-N-(2-hydroxy-2-methylpropyl)-N-methylpyridazine-3-carboxamide

  • MDL number MFCD21867910
  • Cas Number : 1511492-02-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7436493

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7436493
Availability details for
Quantity
  • Availability
  • Add to Cart