ENAH5802DC6D Enamine

tert-butyl (6S)-6-(hydroxymethyl)-2,2-dimethylmorpholine-4-carboxylate

  • MDL number MFCD22690358
  • Cas Number : 1400589-80-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7423130

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7423130
Availability details for
Quantity
  • Availability
  • Add to Cart