ENAH5802DC6F Enamine

ethyl 6-bromo-5-hydroxypyridine-3-carboxylate

  • MDL number MFCD28741320
  • Cas Number : 1805573-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7425998

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7425998
Availability details for
Quantity
  • Availability
  • Add to Cart