ENAH5802DC70 Enamine

3-amino-2-(4,4-difluorocyclohexyl)-1,1,1-trifluoropropan-2-ol hydrochloride

  • MDL number MFCD32068795
  • Cas Number : 2375270-96-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7436029

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7436029
Availability details for
Quantity
  • Availability
  • Add to Cart