ENAH5802DC76 Enamine

methyl (2R)-2-(2-aminoacetamido)-3-(4-hydroxyphenyl)propanoate hydrochloride

  • Cas Number : 2375250-57-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7442705

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7442705
Availability details for
Quantity
  • Availability
  • Add to Cart