ENAH5802DC81 Enamine

[3-cyclohexyl-2-(trifluoromethyl)phenyl]methanol

  • Cas Number : 2375261-06-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7441984

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7441984
Availability details for
Quantity
  • Availability
  • Add to Cart