ENAH5802DC82 Enamine

[3-cyclohexyl-5-(trifluoromethyl)phenyl]methanol

  • Cas Number : 2375270-55-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7440952

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7440952
Availability details for
Quantity
  • Availability
  • Add to Cart