ENAH5802DC86 Enamine

6-(prop-2-yn-1-yl)-6-azaspiro[3.4]octane

  • MDL number MFCD32068682
  • Cas Number : 1856473-49-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-3460309

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-3460309
Availability details for
Quantity
  • Availability
  • Add to Cart