ENAH5802DC8B Enamine

[1-(3,5-dichlorophenyl)-3,3-difluorocyclobutyl]methanol

  • Molecular Weight : 267.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1-(3,5-dichlorophenyl)-3,3-difluorocyclobutyl)methanol

Description

Country of Origin UA

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7438879
Availability details for
Quantity
  • Availability
  • Add to Cart