ENAH5802DC8C Enamine

tert-butyl 3-(2-fluoropyridin-3-yl)-3-hydroxyazetidine-1-carboxylate

  • Cas Number : 1609671-90-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6759930

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6759930
Availability details for
Quantity
  • Availability
  • Add to Cart