ENAH5802DC8D Enamine

tert-butyl N-[4-(2-hydroxyethyl)-6-methoxypyridin-3-yl]carbamate

  • MDL number MFCD24466995
  • Cas Number : 727993-72-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177276

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177276
Availability details for
Quantity
  • Availability
  • Add to Cart