ENAH5802DC90 Enamine

(4-bromophenyl)(1,2-thiazol-5-yl)methanol

  • MDL number MFCD11110343
  • Cas Number : 1049730-17-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-3278885

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-3278885
Availability details for
Quantity
  • Availability
  • Add to Cart