ENAH5802DC97 Enamine

benzyl 1-hydroxy-5H,6H,7H,8H-pyrido[3,4-d]pyridazine-6-carboxylate

  • MDL number MFCD32069190
  • Cas Number : 2375273-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7438810

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7438810
Availability details for
Quantity
  • Availability
  • Add to Cart