ENAH5802DC9B Enamine

3,6-dibromopyrazolo[1,5-a]pyrimidin-2-ol hydrobromide

  • MDL number MFCD32066069
  • Cas Number : 2361644-48-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7432362

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7432362
Availability details for
Quantity
  • Availability
  • Add to Cart