ENAH5802DC9C Enamine

N-(but-3-yn-1-yl)cyclohexanamine hydrochloride

  • MDL number MFCD32069107
  • Cas Number : 113738-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 197-198
Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7438199

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7438199
Availability details for
Quantity
  • Availability
  • Add to Cart