ENAH5802DCA0 Enamine

{5-bromo-2-[methyl(2,2,2-trifluoroethyl)amino]pyridin-3-yl}methanol

  • MDL number MFCD16159555
  • Cas Number : 1250558-07-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7441067

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7441067
Availability details for
Quantity
  • Availability
  • Add to Cart