ENAH5802DCA4 Enamine

2,2,2-trifluoro-1-[3-imino-2-(propan-2-yl)-2,3-dihydro-1H-pyrazol-4-yl]-1-(1-methyl-1H-imidazol-2-yl)ethan-1-ol

  • MDL number MFCD26143536
  • Cas Number : 1955506-87-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25533

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-25533
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart