ENAH5802DCA8 Enamine

ethyl 8-bromo-6-fluoro-4-hydroxyquinoline-3-carboxylate

  • MDL number MFCD09861307
  • Cas Number : 1019015-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7443416

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7443416
Availability details for
Quantity
  • Availability
  • Add to Cart