ENAH5802DCAA Enamine

1-[(2-methoxyphenyl)amino]-3-(naphthalen-1-yloxy)propan-2-ol

  • MDL number MFCD02344635
  • Cas Number : 450363-95-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7355300

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7355300
Availability details for
Quantity
  • Availability
  • Add to Cart