ENAH5802DCCD Enamine

(2E)-4-methylpent-2-enal

  • MDL number MFCD00800501
  • Cas Number : 24502-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1692335

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1692335
Availability details for
Quantity
  • Availability
  • Add to Cart