ENAH5802DCD0 Enamine

5-methylhex-4-enal

  • MDL number MFCD19300697
  • Cas Number : 764-32-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2068469

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2068469
Availability details for
Quantity
  • Availability
  • Add to Cart