ENAH5802DCD3 Enamine

2-ethyl-2-formylbutanenitrile

  • Cas Number : 2344681-57-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7425571

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7425571
Availability details for
Quantity
  • Availability
  • Add to Cart