ENAH5802DCD4 Enamine

3-methoxy-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD09258814
  • Cas Number : 1215632-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6497857

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6497857
Availability details for
Quantity
  • Availability
  • Add to Cart