ENAH5802DCD8 Enamine

2,6-dimethylpyrimidine-4-carbaldehyde

  • MDL number MFCD10696982
  • Cas Number : 1074-39-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1704204

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1704204
Availability details for
Quantity
  • Availability
  • Add to Cart