ENAH5802DCDF Enamine

5-ethyl-1-methyl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD08700635
  • Cas Number : 1007541-44-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2478119

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2478119
Availability details for
Quantity
  • Availability
  • Add to Cart