ENAH5802DCE1 Enamine

5-fluoro-3-methylpyridine-2-carbaldehyde

  • MDL number MFCD18256737
  • Cas Number : 780801-57-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2485211

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2485211
Availability details for
Quantity
  • Availability
  • Add to Cart