ENAH5802DCE3 Enamine

5-(methoxymethyl)-1,3-oxazole-4-carbaldehyde

  • MDL number MFCD28604935
  • Cas Number : 1554168-44-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6931707

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6931707
Availability details for
Quantity
  • Availability
  • Add to Cart