ENAH5802DCE5 Enamine

(2E)-4-[methyl(prop-2-yn-1-yl)amino]but-2-enoic acid

  • Cas Number : 1867088-08-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6760254

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6760254
Availability details for
Quantity
  • Availability
  • Add to Cart