ENAH5802DCE9 Enamine

1-(difluoromethyl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD13188379
  • Cas Number : 1216244-84-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-261992

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-261992
Availability details for
Quantity
  • Availability
  • Add to Cart