ENAH5802DCEA Enamine

5-(difluoromethyl)-1H-pyrazole-3-carbaldehyde

  • MDL number MFCD28655562
  • Cas Number : 2097855-39-3
  • Molecular Weight : 146.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(difluoromethyl)-1H-pyrazole-3-carbaldehyde
Inchi Code 1S/C5H4F2N2O/c6-5(7)4-1-3(2-10)8-9-4/h1-2,5H,(H,8,9)
Inchi Key SSASNWWZWLGETK-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7062654

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7062654
Availability details for
Quantity
  • Availability
  • Add to Cart