ENAH5802DCED Enamine

3-(difluoromethyl)-1,2-oxazole-5-carbaldehyde

  • Cas Number : 2344685-03-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6102173

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6102173
Availability details for
Quantity
  • Availability
  • Add to Cart