ENAH5802DCF1 Enamine

5-cyclobutyl-1,3-oxazole-4-carbaldehyde

  • Cas Number : 2344681-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7179425

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7179425
Availability details for
Quantity
  • Availability
  • Add to Cart