ENAH5802DCF6 Enamine

1-cyclopropylpiperidine-4-carbaldehyde

  • MDL number MFCD28629930
  • Cas Number : 1556703-59-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6793619

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6793619
Availability details for
Quantity
  • Availability
  • Add to Cart