ENAH5802DCF7 Enamine

4-chloro-6-methylpyridine-3-carbaldehyde

  • MDL number MFCD13188650
  • Cas Number : 1060805-86-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008710

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008710
Availability details for
Quantity
  • Availability
  • Add to Cart