ENAH5802DD00 Enamine

2,5-dimethyl-1-(prop-2-yn-1-yl)-1H-pyrrole-3-carbaldehyde

  • MDL number MFCD12090918
  • Cas Number : 1156750-40-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1261834

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1261834
Availability details for
Quantity
  • Availability
  • Add to Cart