ENAH5802DD01 Enamine

4-(difluoromethyl)thiophene-2-carbaldehyde

  • Cas Number : 2375259-38-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7194555

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7194555
Availability details for
Quantity
  • Availability
  • Add to Cart